Please login to your online account to display your discounted pricing

anti-NOD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056878SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056878T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 50% glycerol
Gene ID (Entrez) 10392
Cross Reactivity Human
Primary or Secondary Primary
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Antiserum
Immunogen Amino acids 930-949 of human NOD1 protein were used as the immunogen for this antibody.
Test Specificity Cross reacts with Human. Predicted to cross react with New World Monkey and Mouse.
Conjugate Unlabeled
Gene Accession No. Q9Y239
Antigen NOD1
Regulatory Status RUO
Quantity 0.025mL
Host Species Rabbit