Please login to your online account to display your discounted pricing

anti-NOD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056878

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056878

  • / Each
 

Description & Specifications

Specifications

Antigen NOD1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 50% glycerol
Gene Accession No. Q9Y239
Host Species Rabbit
Immunogen Amino acids 930-949 of human NOD1 protein were used as the immunogen for this antibody.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10392
Test Specificity Cross reacts with Human. Predicted to cross react with New World Monkey and Mouse.
Target Species Human