missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NLRX1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 580-630 of human NLRX1 was used as the immunogen for the antibody.
Antigen NLRX1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227198 Novus Biologicals Canada
NBP227198
0.1mg Each for $329.00
NBP227198SS Novus Biologicals Canada
NBP227198SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
NLRX1
Purified
RUO
The amino acid sequence used as immunogen for the NLRX1 antibody is 100% homologous in human, mouse, rat, monkey and horse, and 95% homologous in cow and dog.
79671
0.5mg/mL
A portion of amino acid 580-630 of human NLRX1 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q86UT6
100μg in 200μL PBS containing 0.05% BSA.