Please login to your online account to display your discounted pricing

anti-NLRP7, Polyclonal, Novus Biologicals™

$109.00 - $274.59

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 106-146 human NALP7 was used as the immunogen
Antigen NLRP7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224556SS

 
novus biologicals canada
NBP224556SS
Unlabeled 0.025mg Each for $109.00

NBP224556

 
novus biologicals canada
NBP224556
Unlabeled 0.1mg Each for $274.59
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 106-146 human NALP7 was used as the immunogen
Antigen NLRP7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and rhesus monkey.
Gene Accession No. Q8WX94
Gene ID (Entrez) 199713
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL