Please login to your online account to display your discounted pricing

anti-NLRP7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224556SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224556SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 199713
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mg
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. Q8WX94
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and rhesus monkey.
Concentration 0.5mg/mL
Antigen NLRP7
Immunogen A synthetic peptide corresponding to amino acids 106-146 human NALP7 was used as the immunogen
Regulatory Status RUO
Cross Reactivity Human
Format Affinity Purified
Host Species Rabbit