Please login to your online account to display your discounted pricing

anti-NLRP7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224556

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224556

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and rhesus monkey.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide corresponding to amino acids 106-146 human NALP7 was used as the immunogen
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Regulatory Status RUO
Gene Accession No. Q8WX94
Gene ID (Entrez) 199713
Conjugate Unlabeled
Cross Reactivity Human
Quantity 0.1mg
Format Affinity Purified
Host Species Rabbit
Antigen NLRP7