missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NLRP2/NALP2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1030-1080 of human NALP2 was used as the immunogen.
Antigen NLRP2/NALP2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224667SS Novus Biologicals Canada
NBP224667SS
0.025mg Each for $109.00
NBP224667 Novus Biologicals Canada
NBP224667
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
NLRP2/NALP2
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms Y5, X9, X3), horse, zebra fish, mouse, chimpanzee, chicken, dog, rat, pufferfish and cow.
55655
0.5mg/mL
A portion of amino acids 1030-1080 of human NALP2 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
Q9NX02
100μg in 200μL PBS containing 0.05% BSA