Please login to your online account to display your discounted pricing

anti-NLRP2/NALP2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human NALP2 was used as the immunogen.
Antigen NLRP2/NALP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224550

 
novus biologicals canada
NBP224550
Unlabeled 0.1mg Each for $255.80

NBP224550SS

 
novus biologicals canada
NBP224550SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human NALP2 was used as the immunogen.
Antigen NLRP2/NALP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a) and chimpanzee, 95% homologous in monkey, horse and cow, 90% homologous in dog and 80% homologous in mouse, rat and xenopus.
Gene Accession No. Q9NX02
Gene ID (Entrez) 55655
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL