Please login to your online account to display your discounted pricing

anti-NLRP2/NALP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224667SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224667SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9NX02
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 1030-1080 of human NALP2 was used as the immunogen.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms Y5, X9, X3), horse, zebra fish, mouse, chimpanzee, chicken, dog, rat, pufferfish and cow.
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 55655
Antigen NLRP2/NALP2
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Quantity 0.025mg
Cross Reactivity Human
Format Purified