Please login to your online account to display your discounted pricing

anti-NLRP2/NALP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224667

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224667

  • / Each
 

Description & Specifications

Specifications

Antigen NLRP2/NALP2
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q9NX02
Host Species Rabbit
Immunogen A portion of amino acids 1030-1080 of human NALP2 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55655
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms Y5, X9, X3), horse, zebra fish, mouse, chimpanzee, chicken, dog, rat, pufferfish and cow.
Target Species Human