Please login to your online account to display your discounted pricing

anti-NLRP10/Pynod/NALP10, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 493-509 of human NALP10 REVQRLLEVKEQEGNDE protein was used as the immunogen.
Antigen NLRP10/Pynod/NALP10
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224606

 
novus biologicals canada
NBP224606
Unlabeled 0.1mg Each for $255.80

NBP224606SS

 
novus biologicals canada
NBP224606SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 493-509 of human NALP10 REVQRLLEVKEQEGNDE protein was used as the immunogen.
Antigen NLRP10/Pynod/NALP10
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 95% homologous in rabbit and 86% homologous in mouse.
Gene Accession No. Q86W26
Gene ID (Entrez) 338322
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL