Please login to your online account to display your discounted pricing

anti-NLRP1/NALP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056147

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056147

  • / Each
 

Description & Specifications

Specifications

Antigen NLRP1/NALP1
Host Species Rabbit
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 22861
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to amino acids 1058-1077 (CVPSPASQGDLHTKPLGTDD) of human NALP1 (NAC/CARD7) was used as immunogen
Test Specificity NALP1 .
Format Antiserum
Quantity 0.05mL
Cross Reactivity Human