Please login to your online account to display your discounted pricing

anti-NKX6.1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149672SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149672SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4825
Gene Accession No. P78426
Primary or Secondary Primary
Formulation PBS, 30% glycerol
Quantity 0.025mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO
Immunogen A genomic peptide made to an internal region of the human Nkx6.1 protein (within residues 50-200). [Swiss-Prot P78426]
Antigen NKX6.1