Please login to your online account to display your discounted pricing

anti-NKX6.1, Polyclonal, Novus Biologicals™

$144.63 - $406.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human Nkx6.1 protein (within residues 50-200). [Swiss-Prot P78426]
Antigen NKX6.1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149672

 
novus biologicals canada
NBP149672
Unlabeled 0.1mL Each for $406.00

NBP149672SS

 
novus biologicals canada
NBP149672SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human Nkx6.1 protein (within residues 50-200). [Swiss-Prot P78426]
Antigen NKX6.1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene Accession No. P78426
Gene ID (Entrez) 4825
Formulation PBS, 30% glycerol
Concentration 0.5mg/mL