missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NKX2.5, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-324 of human NKX2-5 was used as the immunogen.
Antigen NKX2.5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224675SS Novus Biologicals Canada
NBP224675SS
0.025mg Each for $109.00
NBP224675 Novus Biologicals Canada
NBP224675
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
NKX2.5
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, cat, cow, horse, pig and rhesus monkey, 88% homologous in mouse, rat and opossum and 77% homologous in Xenopus and chicken.
1482
0.5mg/mL
A portion of amino acids 300-324 of human NKX2-5 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P52952
100μg in 200μL PBS containing 0.05% BSA