Please login to your online account to display your discounted pricing

anti-NKp80/KLRF1, PE, Clone: 239127, R&D Systems


Manufacturer: r & d systems  RDSFAB1900P025

 View more versions of this product

Catalog No. FAB1900P025

  • / Each
 

Description & Specifications

Specifications

Antigen NKp80/KLRF1
Clone 239127
Conjugate PE
Cross Reactivity Human
Format Purified
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1. Val66-Tyr231
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human