Please login to your online account to display your discounted pricing

anti-NKp80/KLRF1, APC, Clone: 239127, R&D Systems


Manufacturer: r & d systems  RDSFAB1900A025

 View more versions of this product

Catalog No. FAB1900A025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Clone 239127
Cross Reactivity Human
Quantity 25 Tests
Antigen NKp80/KLRF1
Isotype IgG2a
Host Species Rat
Primary or Secondary Primary
Conjugate APC
Monoclonal or Polyclonal Monoclonal
Format Purified
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1. Val66-Tyr231
Purification Method Protein A or G purified from hybridoma culture supernatant