Please login to your online account to display your discounted pricing

anti-NKp46/NCR1, Fluorescein, Clone: 195314, R&D Systems


Manufacturer: r & d systems  RDSFAB1850F025

 View more versions of this product

Catalog No. FAB1850F025

  • / Each
 

Description & Specifications

Specifications

Antigen NKp46/NCR1
Clone 195314
Conjugate Fluorescein
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Mouse T cell hybridoma transfected with human NKp46/NCR1.
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human