Please login to your online account to display your discounted pricing

anti-NKp46/NCR1, APC, Clone: 195314, R&D Systems


Manufacturer: r & d systems  RDSFAB1850A025

 View more versions of this product

Catalog No. FAB1850A025

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Clone 195314
Regulatory Status RUO
Cross Reactivity Human
Antigen NKp46/NCR1
Immunogen Mouse T cell hybridoma transfected with human NKp46/NCR1.
Conjugate APC
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG2b
Format Purified
Quantity 25 Tests
Purification Method Protein A or G purified from hybridoma culture supernatant