Please login to your online account to display your discounted pricing

anti-NKp46/NCR1, APC, Clone: 195314, R&D Systems


Manufacturer: r & d systems  RDSFAB1850A100

 View more versions of this product

Catalog No. FAB1850A100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG2b
Quantity 100 Tests
Clone 195314
Antigen NKp46/NCR1
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp46/NCR1.
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate APC
Cross Reactivity Human