Please login to your online account to display your discounted pricing

anti-NKp30/NCR3, PE, Clone: 210845, R&D Systems


Manufacturer: r & d systems  RDSFAB1849P025

 View more versions of this product

Catalog No. FAB1849P025

  • / Each
 

Description & Specifications

Specifications

Antigen NKp30/NCR3
Immunogen Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera.
Isotype IgG2a
Quantity 25 Tests
Cross Reactivity Human
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 210845
Conjugate PE
Format Purified
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO