Please login to your online account to display your discounted pricing

anti-NKp30/NCR3, PE, Clone: 210845, R&D Systems


Manufacturer: r & d systems  RDSFAB1849P100

 View more versions of this product

Catalog No. FAB1849P100

  • / Each
 

Description & Specifications

Specifications

Antigen NKp30/NCR3
Format Purified
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera.
Isotype IgG2a
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 210845
Conjugate PE
Purification Method Protein A or G purified from hybridoma culture supernatant
Cross Reactivity Human