Please login to your online account to display your discounted pricing

anti-NKp30/NCR3, APC, Clone: 210845, R&D Systems

$139.08 - $432.57

Specifications

Antigen NKp30/NCR3
Clone 210845
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1849A100

 
r & d systems
RDSFAB1849A100
APC 100 Tests Each for $432.57

FAB1849A025

 
r & d systems
RDSFAB1849A025
APC 25 Tests Each for $139.08
Description & Specifications

Specifications

Antigen NKp30/NCR3
Clone 210845
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry