Please login to your online account to display your discounted pricing

anti-NKp30/NCR3, Alexa Fluor 488, Clone: 210845, R&D Systems


Manufacturer: r & d systems  RDSFAB1849G025

 View more versions of this product

Catalog No. FAB1849G025

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG2a
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera.
Quantity 25 Tests
Clone 210845
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Alexa Fluor 488
Antigen NKp30/NCR3
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Cross Reactivity Human