Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, PE, Clone: 191004, R&D Systems

$126.32 - $343.25

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1547P100

 
r & d systems
RDSFAB1547P100
PE 100 Tests Each for $343.25

FAB1547P025

 
r & d systems
RDSFAB1547P025
PE 25 Tests Each for $126.32
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKG2D. Phe94-Val232
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry