Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, Fluorescein, Clone: 191004, R&D Systems

$129.05 - $285.48

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1547F025

 
r & d systems
RDSFAB1547F025
Fluorescein 25 Tests Each for $129.05

FAB1547F100

 
r & d systems
RDSFAB1547F100
Fluorescein 100 Tests Each for $285.48
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKG2D. Phe94-Val232
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry