Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, APC, Clone: 191004, R&D Systems


Manufacturer: r & d systems  RDSFAB1547A100

 View more versions of this product

Catalog No. FAB1547A100

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKG2D. Phe94-Val232
Regulatory Status RUO
Clone 191004
Antigen NKG2D/CD314
Format Purified
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate APC
Cross Reactivity Mouse