Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, APC, Clone: 191004, R&D Systems

$129.05 - $402.80

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1547A025

 
r & d systems
RDSFAB1547A025
APC 25 Tests Each for $129.05

FAB1547A100

 
r & d systems
RDSFAB1547A100
APC 100 Tests Each for $402.80
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone 191004
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKG2D. Phe94-Val232
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry