Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, APC, Clone: 149810, R&D Systems


Manufacturer: r & d systems  RDSFAB139A025

 View more versions of this product

Catalog No. FAB139A025

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Clone 149810
Host Species Mouse
Regulatory Status RUO
Antigen NKG2D/CD314
Conjugate APC
Primary or Secondary Primary
Isotype IgG1
Quantity 25 Tests
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Purification Method Protein A or G purified from hybridoma culture supernatant