Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, APC, Clone: 149810, R&D Systems

$139.08 - $407.05

Specifications

Antigen NKG2D/CD314
Clone 149810
Cross Reactivity Human
Format Purified
Host Species Murine
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB139A025

 
r & d systems
RDSFAB139A025
Allophycocyanin 25 Tests Each for $139.08

FAB139A100

 
r & d systems
RDSFAB139A100
Allophycocyanin 100 Tests Each for $407.05
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone 149810
Cross Reactivity Human
Format Purified
Host Species Murine
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry