Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, Alexa Fluor 700, Clone: 149810, R&D Systems


Manufacturer: r & d systems  RDSFAB139N025

 View more versions of this product

Catalog No. FAB139N025

  • / Each
 

Description & Specifications

Specifications

Purification Method Protein A or G purified from hybridoma culture supernatant
Format Purified
Quantity 25 Tests
Regulatory Status RUO
Cross Reactivity Human
Clone 149810
Conjugate Alexa Fluor 700
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Antigen NKG2D/CD314
Host Species Mouse
Isotype IgG1