Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, Alexa Fluor 700, Clone: 149810, R&D Systems

$142.08 - $389.76

Specifications

Antigen NKG2D/CD314
Clone 149810
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB139N025

 
r & d systems
RDSFAB139N025
Alexa Fluor 700 25 Tests Each for $142.08

FAB139N100

 
r & d systems
RDSFAB139N100
Alexa Fluor 700 100 Tests Each for $389.76
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone 149810
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry