Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, Alexa Fluor 700, Clone: 149810, R&D Systems


Manufacturer: r & d systems  RDSFAB139N100

 View more versions of this product

Catalog No. FAB139N100

  • / Each
 

Description & Specifications

Specifications

Antigen NKG2D/CD314
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 149810
Conjugate Alexa Fluor 700
Cross Reactivity Human