Please login to your online account to display your discounted pricing

anti-NKG2D/CD314, Alexa Fluor 488, Clone: 149810, R&D Systems


Manufacturer: r & d systems  RDSFAB139G100

 View more versions of this product

Catalog No. FAB139G100

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Isotype IgG1
Clone 149810
Antigen NKG2D/CD314
Regulatory Status RUO
Format Purified
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2D/CD314.
Quantity 100 Tests
Primary or Secondary Primary
Conjugate Alexa Fluor 488
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant