Please login to your online account to display your discounted pricing

anti-NKG2C/CD159c, PE, Clone: 134591, R&D Systems


Manufacturer: r & d systems  RDSFAB138P025

 View more versions of this product

Catalog No. FAB138P025

  • / Each
 

Description & Specifications

Specifications

Antigen NKG2C/CD159c
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94.
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 134591
Conjugate PE
Cross Reactivity Human