Please login to your online account to display your discounted pricing

anti-NKG2C/CD159c, PE, Clone: 134591, R&D Systems


Manufacturer: r & d systems  RDSFAB138P100

 View more versions of this product

Catalog No. FAB138P100

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Format Purified
Clone 134591
Cross Reactivity Human
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94.
Conjugate PE
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen NKG2C/CD159c
Isotype IgG1
Quantity 100 Tests
Purification Method Protein A or G purified from hybridoma culture supernatant