Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, PerCP, Clone: 131411, R&D Systems


Manufacturer: r & d systems  RDSFAB1059C100

 View more versions of this product

Catalog No. FAB1059C100

  • / Each
 

Description & Specifications

Specifications

Quantity 100 Tests
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Clone 131411
Cross Reactivity Human
Isotype IgG2a
Host Species Mouse
Antigen NKG2A/CD159a
Primary or Secondary Primary
Conjugate PerCP
Monoclonal or Polyclonal Monoclonal
Purification Method Protein A or G purified from hybridoma culture supernatant
Format Purified
Regulatory Status RUO