Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, PE, Clone: 705829, R&D Systems


Manufacturer: r & d systems  RDSFAB6867P100

 View more versions of this product

Catalog No. FAB6867P100

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2b
Cross Reactivity Mouse
Antigen NKG2A/CD159a
Purification Method Protein A or G purified from hybridoma culture supernatant
Format Purified
Regulatory Status RUO
Clone 705829
Conjugate PE
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Quantity 100 Tests
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a.