Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, PE, Clone: 705829, R&D Systems


Manufacturer: r & d systems  RDSFAB6867P025

 View more versions of this product

Catalog No. FAB6867P025

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a.
Regulatory Status RUO
Clone 705829
Cross Reactivity Mouse
Antigen NKG2A/CD159a
Format Purified
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PE