Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, PE, Clone: 131411, R&D Systems


Manufacturer: r & d systems  RDSFAB1059P100

 View more versions of this product

Catalog No. FAB1059P100

  • / Each
 

Description & Specifications

Specifications

Antigen NKG2A/CD159a
Clone 131411
Conjugate PE
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human