Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, APC, Clone: 705829, R&D Systems


Manufacturer: r & d systems  RDSFAB6867A100

 View more versions of this product

Catalog No. FAB6867A100

  • / Each
 

Description & Specifications

Specifications

Antigen NKG2A/CD159a
Clone 705829
Conjugate Allophycocyanin
Cross Reactivity Mouse
Format Purified
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a.
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Murine