Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, APC, Clone: 131411, R&D Systems


Manufacturer: r & d systems  RDSFAB1059A100

 View more versions of this product

Catalog No. FAB1059A100

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a
Antigen NKG2A/CD159a
Format Purified
Regulatory Status RUO
Quantity 100 Tests
Primary or Secondary Primary
Conjugate APC
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Clone 131411
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Purification Method Protein A or G purified from hybridoma culture supernatant
Cross Reactivity Human