Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, Alexa Fluor 700, Clone: 131411, R&D Systems


Manufacturer: r & d systems  RDSFAB1059N100

 View more versions of this product

Catalog No. FAB1059N100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Quantity 100 Tests
Cross Reactivity Human
Antigen NKG2A/CD159a
Host Species Mouse
Isotype IgG2a
Regulatory Status RUO
Primary or Secondary Primary
Clone 131411
Conjugate Alexa Fluor 700
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal