Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, Alexa Fluor 700, Clone: 131411, R&D Systems

$177.37 - $432.57

Specifications

Antigen NKG2A/CD159a
Clone 131411
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1059N025

 
r & d systems
RDSFAB1059N025
Alexa Fluor 700 25 Tests Each for $177.37

FAB1059N100

 
r & d systems
RDSFAB1059N100
Alexa Fluor 700 100 Tests Each for $432.57
Description & Specifications

Specifications

Antigen NKG2A/CD159a
Clone 131411
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry