Please login to your online account to display your discounted pricing

anti-NKG2A/CD159a, Alexa Fluor 700, Clone: 131411, R&D Systems


Manufacturer: r & d systems  RDSFAB1059N025

 View more versions of this product

Catalog No. FAB1059N025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen NKG2A/CD159a
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94.
Isotype IgG2a
Primary or Secondary Primary
Quantity 25 Tests
Monoclonal or Polyclonal Monoclonal
Clone 131411
Conjugate Alexa Fluor 700
Cross Reactivity Human
Purification Method Protein A or G purified from hybridoma culture supernatant