Please login to your online account to display your discounted pricing

anti-NK1R, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor [UniProt No. P25103]
Antigen NK1R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300119

 
novus biologicals canada
NB300119
Unlabeled 0.1mL Each for $384.00

NB300119SS

 
novus biologicals canada
NB300119SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor [UniProt No. P25103]
Antigen NK1R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Reacts with human, mouse and rat. Immunogen has 94% sequence identity with guinea pig. Mouse reactivity is from a customer review.
Gene Accession No. P25103
Gene ID (Entrez) 6869
Formulation PBS, 30% glycerol
Concentration 1.0mg/mL