Please login to your online account to display your discounted pricing

anti-NIFK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056661

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056661

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 84365
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9BYG3
Regulatory Status RUO
Host Species Rabbit
Applications Western Blotting
Test Specificity In NIH 3T3, a 34 kDa band is observed.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antiserum
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 75-90 of mouse NIFK.
Monoclonal or Polyclonal Polyclonal
Antigen NIFK
Quantity 0.2mL