Please login to your online account to display your discounted pricing

anti-Niemann-Pick type C1 Like-1, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 500-600). [UniProt No. Q6T3U3]
Antigen Niemann-Pick type C1 Like-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400128

 
novus biologicals canada
NB400128
Unlabeled 0.1mL Each for $362.00

NB400128SS

 
novus biologicals canada
NB400128SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 500-600). [UniProt No. Q6T3U3]
Antigen Niemann-Pick type C1 Like-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 29881
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL