Please login to your online account to display your discounted pricing

anti-Niemann-Pick type C1 Like-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB400128

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB400128

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation Tris-citrate/phosphate, pH 7-8
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Niemann-Pick type C1 Like-1
Immunogen A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 500-600). [UniProt No. Q6T3U3]
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 29881
Concentration 1mg/mL
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL
Host Species Rabbit