Please login to your online account to display your discounted pricing

anti-NFkB1/NFkB p105, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177395

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177395

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene ID (Entrez) 4790
Gene Accession No. P19838
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Concentration 1.18mg/mL
Antigen NFkB1/NFkB p105
Format Affinity Purified
Regulatory Status RUO
Immunogen A genomic peptide made to an N-terminal region of the human NFkB p105/p50 protein (within residues 400-590). [Swiss-Prot P19838]
Quantity 0.1mL